Would you like to log in again?
Cleanroom Paper and Sticky Pads
Paper & Sticky Pads
Cleanroom paper has a smooth surface (latex impregnated) with minimal particles and ions and this reduces possible contamination in cleanrooms and controlled environments. The cleanroom paper is lightly coloured for easy differentiation from normal paper. It accepts writing, copying and printing on all common office equipment and has an excellent retention of toners /inks.
Sticky pads are made of cleanroom paper. Cleaned, packed in a class ISO 5 cleanroom. Cleanroom sticky pads contain removable water-based solvent-free adhesive with very low outgassing levels. They can be stuck and removed without leaving residue on the support. For annotating documents or leaving messages in all cleanroom environments. Our sticky pads are available in different colours.
Paper in different colours
Sticky Pads