Would you like to log in again?
Tyvek Cleanroom Packaging
Tyvek Packaging
Features:
  • Composite of polyethylene foil and uncoated Tyvek®
  • Transparent foil offers the advantage of product visibility
  • Steam-sterilisable up to 125 °C
  • Ethylene oxide- and gamma sterilisable
  • Very low gas residue (for instance ETO) gas sterilisation shortens the airing out time
  • Sterilisation indicators on the packaging
  • Extremely low particle contamination
  • Outstanding barrier characteristics
  • High-quality sealing and burst strength
  • Easily heat-sealed (sealing temperature up to max. 120 °C)
  • Foils are suitable for printing
  • 100% recyclable
  • Optionally available as flat bag or easy-tear-bag to minimise particle contamination
  • Available in various cleanroom classes
Easy-tear Bag