Would you like to log in again?
Polyethylene Cleanroom Packaging
Polyethylene Packaging
Features:
  • Ultra-clean, clear polyethylene
  • Transparent foil offers the advantage of product visibility
  • Suitable for gamma sterilisation
  • Sterilisation indicators on the packaging
  • No gas emissions
  • Resistant to most chemicals
  • Easily heat-sealed (sealing temperature 120 °C to 200 °C)
  • Optionally available as flat bag or tube
  • Certificate of conformity with each delivery
Flat Bag:
  • Various widths and lengths
  • Standard thickness: 0.100 mm
Tube:
  • Various widths and lengths
  • Standard thickness: 0.100 mm
  • Tube with plastic core, the ideal packaging for oversized sterilised goods