Would you like to log in again?
Anti-static Polyethylene Packaging
Anti-static Polyethylene Packaging
Features:
  • Ultra-clean polyethylene in pink
  • Non-corrosive
  • Resistant to most chemicals
  • Optionally available as flat bag or tube
  • Easily heat-sealed (sealing temperature 120 °C to 200 °C)
  • Certificate of conformity with each delivery
 Flat bag:
  • Various widths and lengths
  • Standard thickness: 0.100 mm
  • Optionally available with self-adhesive seal
Tube:
  • Various widths and lengths
  • Standard thickness: 0.150 mm
  • The ideal packaging for oversized sterilised goods
  • Tube with plastic core