Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Cleanroom Notebooks
Cleanroom notebooks are constructed with cleanroom paper, processed and packed in a ISO class 5 environments. Cleanroom notebooks are the ideal tool for note taking in cleanrooms and controlled environments.
The spiral-bound covers are made from clear antistatic polypropylene, with no amides or silicon. The inks used for printing have low levels of sodium, to reduce ionic contamination. The ruled white paper is opaque enough to allow writing on both sides.
Our recommendation: Cleanroom pens