Would you like to log in again?
Sit-over Cleanroom Benches
Sit-over Benches
Sit-over benches are a functional barrier to separate different areas. They allow a controlled and low-particle entry and leave of employees working in a cleanroom or in controlled environments. Walking shoes are stored in the compartments provided in the sit-over bench, then the cleanroom personnel swings the legs over on the "clean" side.
Cleanroom compatible with stainless steel sitting surfaces, optionally in polyethylene or polypropylene, with or without shoe compartments. 
Places are numbered to aid allocation of seats to personnel. Available with melamine faced chipboard and/or special design.