Would you like to log in again?
Dew-point and humidity meters
Over time, a humidity meter is subject to fluctuations in its accuracy of measurement. This period depends on the way in which it is used in the cleanroom or controlled environment. To be able to guarantee the accuracy and to maintain it within acceptable limits, the device must be calibrated at regular intervals.
Humidity Instruments
For certain specific applications in the pharmaceutical industry, the calibration must be carried out every 3 months, as measuring devices are subject to the validation process and even the smallest fluctuations can have a large influence on the application.
Humidity calibrator
  • For calibration of humidity sensors, transmitters and portable devices
  • Generates stable humidity values of 10 to 90% RH
  • Compact and easy to transport
  • Rapid stabilisation of the test chamber
  • Optional: independent traceable reference hygrometer
  • Several sensors can be calibrated simultaneously
Humidity / temperature calibrator
  • For calibration of rF humidity sensors and transmitters
  • Temperatures from 10 to 50 °C with a precision of 0.1 °C and humidity of 10 to 90% RH
  • Precise and repeatable calibrations are possible
  • Generates humidity and temperature simultaneously
  • Excellent stability
  • Compact and easy to transport
  • Optional: built-in data recorder, logs the built-in reference sensor and the sensors to be calibrated
  • Suitable for sensors with Ø betw. 5 and 25 mm
Contact us
Our recommendation:
Expert calibration of the devices is necessary. In normal environments, one calibration per year is sufficient.