Would you like to log in again?
Dry Storage Systems
Moisture sensitive components (those tending to corrode within normal atmosphere) or components which have to be protected from particles, should be stored within a dry atmosphere.
Dry Storage Systems
For this purpose dry storage systems can be used. Within these systems the stored goods are surrounded by a dry and inert atmosphere, if necessary. To achieve the stable and dry atmosphere there are 3 different technologies of dehumidification available.
  • Nitrogen dehumidification: controlled feed-in of nitrogen
  • Adsorption dehumidification: controlled drying of the air with a high efficient adsorption dehumidifier
  • Compressed-air dehumidification: controlled feed-in of compressed air
Features:
  • Materials: transparent acrylic glass or powder coated steel
  • For white light- sensitive components: design acrylic yellow (AMBER) available
  • Control unit with automatic flushing (system flushes only when needed)
  • ESD version (resistance 107 Ohm/cm²)
  • Optimal visual and acoustic alarms
  • Optional chamber system (division of the cabinet volume in individual chambers)
  • Optional with lockable doors
  • Optional data logging functions
  • Many more customised interior designs of the systems
Contact us
Customized Solutions
Apart from the proven standard products we also provide custom made solutions that can be adapted to any requirement.