Would you like to log in again?
Cleanroom Cabinets
Cabinets
Made of stainless steel, brushed or electropolished. With hinged or sliding doors (lockable doors are optionally available), internal shelves as required, solid or perforated (hanging rail and clothes hangers are optionally available).
Available with melamine formaldehyde resin coating splint disc and with extra equipment. Further extra equipment and customised dimensions are available.