Would you like to log in again?
Disposable Cleanroom Sleeve Protectors
Sleeve protectors should be worn in addition to aprons or when working with isolators or other devices, where the protection of the arms is sufficient.
Sleeve Protectors They have an elastic adjustment on both sides and are available in different lengths, materials and colours. Chemotherapy protective sleeves offer a maximum comfort and protection, recommended use in cleanroom class A (ISO 4). Chemotherapy protective sleeves are antatomical formed with elastic adjustment for optimal fit. Available as sterile and non sterile versions.
Our recommendation
Dispenser systems for the simple collection of disposables such as sleeve protectors, hoods, facemasks, shoe covers or gloves.