Would you like to log in again?
Disposable Overalls
Overalls
Disposable overalls protect sensitive products as well as the operators. They are manufactured specifically to prevent particle contamination and static build up. 
Disposacle cleanroom overalls are made from different materials (Tyvek, Polypropylene, breathable SMS fabric, Polyethylene, Non woven, clean tough fabric).
Sterile and non sterile versions of disposable cleanroom overalls with elasticated wrists and ankles are available.
Upon request with integrated hood. 
Upon request
Single hoods are available.