Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Dispenser for Cleanroom Wipes
Dispenser for Wipes
Dispenser for cleanroom wipes are ideal for a clean storage of your wipe assortment and suitable for table dispensing (available in different sizes).
basan chempump flasks assist with the dosing and storage of volatile solutions. They have a specially designed closure for economical use of the liquid and to avoid evaporation losses after removal of liquids.