Would you like to log in again?
Sterile Solutions for Surfaces in Cleanrooms
In Grade A and B cleanrooms, the disinfectants and detergents must be sterile. Our products are formulated and manufactured according to GMP standards. They are available with extensive technical documentation and supplied with certificates of analysis and sterility.
Surfaces - sterile
The solutions are filtered to 0.2 µm before filling and packaging in cleanroom conditions, and are either aseptically filled or terminally irradiated.  
Neutral detergent
  • High-foaming liquid detergent, neutral pH
  • 0.2 µm filtered, gamma irradiated, double bagged
  • Supplied with certificates of analysis, irradiation and sterility
  • Supplied as: 100 x 30 ml or 50 x 50 ml concentrate
Ethanol and IPA
  • Broad spectrum disinfectant
  • Passes CEN 1276, CEN 1650 and CEN 13697
  • 0.2 µm filtered, gamma irradiated (900 ml) or aseptically filled (5 litres), double bagged
  • Supplied with certificates of analysis, sterility and irradiation, where applicable
  • Denatured Ethanol: 70% ethanol, 30% water (WFI) denatured with IPA
  • Isopropyl alcohol: 70% isopropyl alcohol, 30% water (WFI)
  • Also available as an aerosol with purified water
  • Available as: 6 x 900 ml spray bottle; 6 x 1 l spray bottle; 4 x 5 l or 2 x 5 l canister; 12 x 300 ml aerosol
Sporicidal disinfectants
  • Two-part chlorine dioxide generating system
  • Passes CEN 1276, CEN 1650, CEN 13697 and CEN 13704
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Based on chlorine dioxide
  • Compatible in rotation with amphoteric disinfectants
  • Effective against spores within only 5-8 minutes!
  • Available as: 25 x (2 x 100 ml) concentrate;    
    4 x (4.9 l + 100 ml) ready-to-use
Amphoteric disinfectants
  • Highly effective surface active disinfectant
  • Passes CEN 1276, CEN 1650 and CEN 13697
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Validated HPLC and Titration test methods available
  • Compatible in rotation with a sporicidal or QAC disinfectant
  • Available as: 50 x 50 ml or 50 x 100 ml concentrate;
    6 x 900 ml spray bottle; 4 x 5 l or 2 x 5 l canister  
Quaternary Ammonium disinfectants
  • Highly effective surface active disinfectant based on Quats
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Highly compatible with materials
  • For a broad range of applications
  • Unit: 50 x 25 ml or 50 x 100 ml concentrate;
    6 x 900 ml spray bottle; 4 x 5 l or 2 x 5 l canister
Contact us
The solutions are usually available in various forms:
In concentrate shot-bottles, ready-to-use spray bottles and ready-to-use 5 l canisters.
The spray bottles feature anti-suck technology in order to avoid contaminating the solution with outside air. It can either be a fully sealed system, or a 0.2 µm filtered system.
Both technologies feature similar performance, and allow the content to remain sterile for several weeks after opening.