Would you like to log in again?
Cleanroom Clipboards
Clipboards
Clipboards are the ideal portable writing surface for use of single sheets in cleanrooms and critical environments.
Securely holds 1–50 sheets of cleanroom paper. The clipboards are constructed of rigid polypropylene with a durable plastic clip mechanism for long-lasting use.
We offer a wide range of cleanroom paper that fit the clipboards: