Asecos Safety Cabinets
The different types of safety cabinet
Q-Line: Safety cabinets for flammables
Q-Line safety cabinets store all your flammable products. For example: Acetone, dichloromethane, cyclohexane, diethyl ether, ethanol, hexane, xylene, propanol, etc.
UB-Line: Cooled underbench cabinet UB-S-90K
These under-bench safety cabinets store flammable liquids. UB-Line cabinets can fit under your hood or fume cupboard. Available also refrigerated and filtered ventilation.
V-Line: The space-saver for the laboratory and industry
This security cabinet offers the same storage volume as a standard cabinet. V-Line cabinets bring more convenience with vertical drawers for easy product handling and storage.
SL-Line: Safety cabinets for acids and alkalis
SL-Line cabinets can store your corrosive products: Nitric acid, hydrochloric acid, sodium hydroxide, potassium hydroxide, etc.
FREE guide on the risks of storing and handling hazardous chemicals
The new edition of the asecos® Hazmat Guide contains a wide range of expert information with regards to professional storage and handling of hazardous materials: Including terms and definitions, different types of storage, advantages of a fire resistant safety storage cabinet, technical ventilation and much more…
Do you want to store hazardous substances safely?
asecos® has suitable safety storage cabinets for every application.
Use our configurator to create your individual solution.
Check the performance of asecos® safety cabinets by watching these short videos
Safety storage cabinets with fully automated vertical drawers- brand new from asecos®.
BE SAFE! Safety storage under-bench cabinet with refrigerating unit and recirculating air filter system for the unrestricted storage of flammable hazardous materials in working areas, according to EN 14470-1 and TRGS 510 with a fire resistance of 90 (Type 90) minutes.
In this impressive video you can see a comparison of actually used cabinet types within a stress test – LOOK and THINK- it`s about your personal SAFETY!
What makes a storage cabinet safe? Explore the characteristics that makes sure you make the right decision.
Please have a browse through the ASECOS Highlights- and ask our Experts for YOUR individual solution!
Negligence during the storage of hazardous materials can have serious consequences
In case of fire…
The storage of flammable products will not only cause a rapid spread of fire but also explosions
This will cause injuries and death
Damage to the environment
Destruction of premises and cessation of activities
Legal consequences
And insurance companies will refuse to pay any compensation