Would you like to log in again?
Cell Proliferation Assay Kit XTT
Cell Proliferation Assay Kit XTT
Cell proliferation assays are widely used in Cell biology for the study of growth factors, cytokines or media components.
They are also applied in the screening of cytotoxic agents and lymphocyte activation in order to determine the number of viable cells.
Applichem's Cell Proliferation Kit XTT makes use of XTT as only the mitochondria of living cells are capable to reduce it to an orange-coloured water soluble dye which can be measured and is proportional to the number of metabolically active cells.