Would you like to log in again?
MICROGARD® 2000
MICROGARD® 2000
VALID UNTIL: 30/11/2017
Comfort and exceptional overall barrier protection
The MICROGARD® 2000 standard provides an exceptional overall barrier to low hazard chemicals, particles and biological agents. Ideal for a wide range of industrial applications.
  • Protection - excellent liquid penetration resistance and barrier to fine particulates
  • Comfort - moisture vapour permeable (breathable) to help reduce the risk of heat stress
  • Silicone-free – critical in spray painting applications
  • Ultra-low linting – reduced risk of contamination in critical areas
  • Standards: Type 5-B, Type 6-B. Anti-static – tested according to EN 1149-5
Colour Clothing size Pk Cat. No. Promo price Quantity
With hood (model 111)
Green S 40 113-8766E
Green M 40 113-8767E
Green L 40 113-8768E
Green XL 40 113-8769E
Green XXL 40 113-8770E
Green 3XL 40 113-8771E
White S 40 113-8772E
White M 40 113-8773E
White L 40 113-8774E
White XL 40 113-8775E
White XXL 40 113-8776E
White 3XL 40 113-8777E
With collar (model 103)
White S 40 113-8778E
White M 40 113-8779E
White L 40 113-8780E
White XL 40 113-8781E
White XXL 40 113-8782E
White 3XL 40 113-8783E
With hood, boots and ant-slip soles (model 122)
White S 30 113-8784E
White M 30 113-8785E
White L 30 113-8786E
White XL 30 113-8787E
White XXL 30 113-8788E
White 3XL 30 113-8789E