Would you like to log in again?
Microflex® 93-260 nitrile and neoprene double-coated disposable gloves
Microflex® 93-260 nitrile and neoprene double-coated disposable gloves
Thinnest chemical resistant synthetic composite disposable glove
  • Three layer design for superior protection against harsh chemicals including acids, solvents and bases
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
  • Lower acceptable pin hole rate (0,65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone-free formulation and processing ensure better product protection
Unique multi-layered disposable gloves provides full chemical protection without compromises meeting the high expectation of users for comfort and dexterity.