Would you like to log in again?
AlphaTec® Isolator Box Gloves from Ansell made with revolutionary EPDM material
AlphaTec® Isolator Box Gloves from Ansell made with revolutionary EPDM material
Increase protection and reduce glove replacements with AlphaTec­® EPDM and EPDM+ Isolator Box Gloves, especially designed for Life Sciences applications
AlphaTec® EPDM and EPDM+ Isolator Box Gloves from Ansell deliver strong protection and ruggedness, without compromising on comfort, dexterity and tactile sensitivity. Unique white EPDM+ with a double-layer breach detection system, for Life Science applications in which visual identification of breaches and contaminants is vital.
  • Excellent quality and consistency, available in two colours: black EPDM and white EPDM+
  • Assured compliance with FDA food contact regulations (FDA CFR21)
  • Suitable for use with a range of chemotherapy drugs
  • Designed for repeated sterilisation - reducing the need for glove replacements
  • Comfortable and dexterous with good tactile sensitivity
In an industry where a small glove breach can considerably impact costs and operations, selecting the right glove for the job is crucial. That's why Ansell now offers an even wider range of box gloves to meet your needs. Our new Isolator Box Gloves, made of CSM, EPDM and EPDM+ materials, are built for optimum comfort, protection and longevity. Safeguard your workers and products and increase productivity.
Size Colour Length Thickness Ø Cat. No. Price Quantity
Medium weight
9,5 Black 800 mm 0,4 mm 203 mm 112-4342E
11 Black 800 mm 0,4 mm 203 mm 112-4341E
9,5 Black 800 mm 0,4 mm 250 mm 112-4467E
11 Black 800 mm 0,4 mm 250 mm 112-4466E
11 Black 800 mm 0,4 mm 300 mm 112-4471E
11 Black 800 mm 0,4 mm 300 mm 112-4470E
Heavy weight
9,5 Black 800 mm 0,6 mm 203 mm 112-4344E
11 Black 800 mm 0,6 mm 203 mm 112-4343E
9,5 Black 800 mm 0,6 mm 250 mm 112-4469E
11 Black 800 mm 0,6 mm 250 mm 112-4468E
9,5 Black 800 mm 0,6 mm 300 mm 112-4473E
11 Black 800 mm 0,6 mm 300 mm 112-4472E
EPDM, with lining
9,5 White 800 mm 0,5 mm 203 mm 112-4475E
11 White 800 mm 0,5 mm 203 mm 112-4474E
9,5 White 800 mm 0,5 mm 250 mm 112-4477E
11 White 800 mm 0,5 mm 250 mm 112-4476E
9,5 White 800 mm 0,5 mm 300 mm 112-4479E
11 White 800 mm 0,5 mm 300 mm 112-4478E