Would you like to log in again?
Amersham™ Nitrocellulose Western blotting membranes Selection guide
Amersham™ Nitrocellulose Western blotting membranes Selection guide
GE Healthcare Life Sciences offers an extensive line of 100% pure cellulose filter paper membranes manufactured entirely from high-quality cotton linter. The membranes are manufactured and tested specifically for chromatographic and blotting applications and contain no additives that can cause contamination during the transfer step. High purity gives the membranes the excellent wicking capability and uniform capillary action that is important for obtaining clean and even transfers during blotting.