Would you like to log in again?
Product Ranges
Product Ranges The ACE® range of silica phases, in addition to a wide range of ultra-pure standard selectivity phases (C18, C8, C4, CN, AQ, Phenyl, Amino and Silica), includes several unique selectivity chemistries and a series of high performance wide pore (300Å) phases. The ACE® Excel™ range of columns is based on high pressure hardware, suitable for UHPLC/HPLC and fast analysis applications up to 1,000 bar (15,000 psi).
Innovation and Choice
ACE products are designed to incorporate the latest developments in stationary phase technology. Innovative phases such as the ACE C18-AR, ACE C18-PFP and ACE C18-Amide are designed to offer the hydrophobic characteristics and ruggedness of a C18 phase with the alternative selectivity offered by a phenyl phase, a pentafluorophenyl phase and an amide phase respectively. This provides chromatographers with more choices for alternative selectivity but without compromising stability or robustness. Other unique products include ACE® SuperC18™, a highly inert, pH stable silica phase and ACE CN-ES, a higher stability CN-like selectivity phase, ideal for working with more polar compounds. The range also includes ACE® UltraCore™ SuperC18™ and ACE® UltraCore™ SuperPhenylHexyl™, solid-core particle columns for high efficiency, low pressure separations across a wide pH spectrum.
Advanced Chromatography Technologies’ products are manufactured at purpose built facilities accredited to ISO 9001:2008 and ISO 14001:2004 standards and are available to chromatography users all over the world.