Would you like to log in again?
Discover the 3M™ Versaflo™ Powered Air Turbo TR-600
Discover the 3M™ Versaflo™ Powered Air Turbo TR-600
VALID UNTIL: 30/11/2017
Simply put: It's designed to help make breathing and working easier
Via research, and an understanding of your needs, we have created the most advanced, versatile solution yet.
  • Comfortable airflow - 3 user-selectable flow rates to maximise comfort
  • Ease of use - no calibration, LED battery/filter indicator, audio vibration alarm
  • Multiple applications - easy clean, suitable for shower decontamination and temporary submersion
  • Safe - approved to EN 12941 Class TH2 or TH3 (dependent on headtop)
Description Pk Cat. No. Promo price Quantity
Versaflo Turbo unit TR-602E 1 111-2218E
Versaflo Turbo TR-619E Starter Kit with accessories 1 111-2220E
TR-6130E, ABE1P filter 5 111-2219E
TR-6600, pre-filter 10 111-2222E
BT-30 breathing tube 1 111-1061E
TR-627 easy clean belt 1 111-2221E
BPK-01 harness for backpack 1 111-2199E
TR-329 suspenders/braces for TR-600 1 111-2217E