Would you like to log in again?
VWR Presents 3M Solutions from Detection to Protection and Validation
As a key strategic partner to VWR in personal protective equipment and safety solutions, 3M is passionate about protecting the health and safety of workers in all types of industries and workforce environments.
3M, as a global authority in helping you solve your most complex hearing protection challenges, has launched great tools and services to assist you – from detection to protection to validation.
Use this new platform to download these new tools and services to complement the VWR offering of personal protection equipment.
Detecting noise levels in the workplace is the first step to ensure your workforce is protected.
Complying with regulations, selecting appropriate protection, and analyzing noise control options are all easier with 3M solutions for exposure assessment, noise analysis, and employee engagement.
3M's accurate, reliable, and flexible instrumentation coupled with powerful software is the innovative solution for your noise detection challenge.
View the latest hearing standards
The European Noise Directive Pocket Guide
Once you have detected your noise level, it is critical to select a hearing protector that your employees are motivated to wear.
3M has the broadest selection of hearing protectors for a wide range of environments. Trouble fitting your plugs? Need to communicate in noise? Dirty hands? Hot, humid conditions?
These are just a few of the many challenges that 3M's innovative products can solve for you.
How to select hearing protective equipment
Use and care of hearing protection
You have offered hearing protection to your workers. Now, how do you gain confidence they are protected?
3M™ EARfit™ Dual-Ear Validation System utilizes F-Mire technology to measure the level of employee hearing protection in just 10 seconds. Workers receive their own personal attenuation rating (PAR) and learn how correctly inserting the right earplug increases their level of protection.
How to improve Field Performance of Hearing Protectors
Hearing Protection, Production Insight Magazine April 2016
Why hearing protection is needed
Fitting Foam Earplugs
Fitting Pre-molded Earplugs
Validation System for Ear Fit Testing
3M™ E-A-R Classic™ Earplugs
One Touch™ Pro Earplugs Dispenser
3M™ Peltor™ X Series Earmuffs
3M Product (click to order in our webshop)
Features Attenuation
in dB (SNR)
3M™ Roll-down ear plugs
3M™ Roll-down ear plugs (corded)
The standard disposable solution for long exposure periods
Metal detectable/
Small ear canals/ Corded version/ Colours/Material
from 28dB to 39dB
3M™ Pod ear plugs
The smart solution for hygienic, low cost hearing protection. No roll down required
Corded version/ Colours
from 28dB to 35dB
3M™ Pre-moulded reusable ear plugs
The practical reusable solution with resealable pack or storage case
Corded/ Metal detectable/ Transparent/ Low attenuation
from 20dB to 35dB
3M™ Earplug Dispenser
The easiest way to get your 3M Ear Plugs ready for use for the users
Banded Ear Plugs
Perfect solution for intermittent wearing periods and limited exposure to noise
Multiposition/ Pod shapes/ Low attenuation
from 23dB to 26dB
3M™ PELTOR™ Earmuffs X Series
All available as headband or helmet mounted version.
Newly designed spacer for improved attenuation.
Helmet mounted
from 26dB to 28dB
Earmuffs, Peltor™ X Series
Extremely slim and lightweight earcups providing excellent compatibility when used with other 3M PPE.
Headband/ Helmet mounted
from 26dB to 28dB
Earmuffs, Peltor™ X Series
The earmuff for extremely high noise environments.
Headband/ Helmet mounted
from 34dB to 35dB