Would you like to log in again?
New 3M™ organic gas and vapour filters with Service Life Indicators
When should I change my filters?
New 3M™ organic gas and vapour filters with Service Life Indicators
VALID UNTIL: 30/11/2017
Service Life Indicator technology to help increase confidence in protection
This unique 3M 'end of service life' indicator is a simple visual tool that can help users determine when to change their filters*
  • Suitable for 3M 6000 and 3M 7000 series half and full face masks
  • Designed to indicate service life based on individual exposure and respiratory use patterns
  • Can help provide added peace of mind as a complement to your current changeout schedule or, in some cases, replace your current practices
  • This technology can help increase compliance with your company’s respiratory policy and industry requirements
  • NOTE: Please see the 6051i User Instructions or the 3M Select and Service Life Software (3M.eu/SLS ) to determine if these filters are appropriate for your work environment
Description Pk Cat. No. Price Quantity
3M Gas & Vapour Filter, Service Life Indicator A2, 6055i 8 111-2190E
3M Gas & Vapour Filter, Service Life Indicator A1, 6051i 8 111-2153E