Would you like to log in again?
Due to a system upgrade on Friday 24th February 2017 there will be a 12 noon cut off for orders required for delivery on Monday 27th February
Laboratory Autoclaves and Accessories
Laboratory Autoclaves and Accessories
Autoclaves are used to sterilise equipment and supplies by subjecting them to high pressure saturated steam.
Typical loads include laboratory glassware, equipment, instruments and of course culture media. A growing area of autoclave usage is the pre-disposal treatment and sterilisation of waste material, such as pathogenic waste before final disposal.
Offering both bench top and the larger laboratory autoclaves, with a choice of either front or top loading, they come in a variety of sizes which are available from all major suppliers as well as our own VWR Collection range.
If you need validation of your autoclaving cycle then VWR also offers a huge portfolio of data loggers as well as other essential accessories such as baskets, trays and foil.