Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Volumetric reagents for water determination by the Karl Fischer method

Supplier: VWR Chemicals
These reagents are especially suitable for water determination in aldehydes and ketones.

  • Fast and stable endpoints for accurate results time and time again
  • Low toxicity and pyridine-free
  • Provide consistent results



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

Volumetric one-component titrating reagents
Description Pk VWR Catalog Number Quantity
CombiNORM 1, 1 mg H2O/ml, used with methanol 1 l 85450.290
CombiNORM 2, 2 mg H2O/ml, used with methanol 1 l 85451.290
CombiNORM 5, 5 mg H2O/ml, used with methanol 1 l 85452.290
CombiNORM 5K, for one-component KF titrations in aldehydes and ketones, 5 mg H2O/ml, used with Solvent K 1 l 85463.290
Combi solvent E (ethanol-based working medium) 1 l 85454.290
Solvent K (working medium), for one-component KF titrations in aldehydes and ketones, used with CombiNORM 5K 1 l 85464.290
Lipo solvent CM, non polar reagent (working medium), for non polar substances, oils and fats 1 l 85455.290
Lipo solvent HM, non polar reagent (working medium), for non polar substances, oils and fats 1 l 85456.290
Methanol quick (working medium), for accelerating one-component KF titration, used with CombiNORM 1 l 85457.290
Keto solvent, working medium, for one-component KF titrations in aldehydes and ketones 1 l 85459.290
CombiNORM 5, 5 mg H2O/ml, used with methanol 2,5 l 85452.320

Volumetric two-component titrating reagents
Description Pk VWR Catalog Number Quantity
TitrANT 1, 1 mg H2O/ml, used with Solvent 1 l 85465.290
TitrANT 2, 2 mg H2O/ml, used with Solvent 1 l 85466.290
TitrANT 2E, ethanol based, 2 mg H2O/ml, used with Solvent E 1 l 85467.290
TitrANT 5, 5 mg H2O/ml, used with Solvent 1 l 85468.290
TitrANT 5E, ethanol based, 5 mg H2O/ml, used with Solvent E 1 l 85469.290
Solvent (working medium), for used with all TitrANTs 1 l 85453.290
Solvent (working medium), for used with all TitrANTs 2,5 l 85453.320
Solvent E, ethanol based working medium 1 l 85460.290
Solvent (crude), working medium, for titration in oils 1 l 85458.290
Solvent OIL (working medium), for titration in oils 1 l 85462.290
Solvent CM (working medium), for titration in oils 1 l 85461.290
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty