Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer solutions, secondary standards

Supplier: VWR Chemicals
  • Solution standardised at 25 °C, compliant with DIN 17025
  • Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
  • Fully traceable for optimum pH calibration (traceable to NIST)
  • Supplied with detailed certificate of analysis
  • Temperature dependence data is printed on the labels as are batch number and expiry date


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
pH value pH accuracy Reference temperature Pk VWR Catalog Number Quantity
1,679 ±0,010 25 °C 500 ml 84580.260
4,005 ±0,010 25 °C 500 ml 84582.260
6,865 ±0,010 25 °C 500 ml 84583.260
7,413 ±0,010 25 °C 500 ml 84584.260
9,18 ±0,010 25 °C 500 ml 84585.260
10,012 ±0,010 25 °C 500 ml 84586.260
12,454 ±0,050 25 °C 500 ml 84587.260
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty