Would you like to log in again?
Lab air quality sensor, Halo Sense
Supplier: Erlab
Halo Sense has been designed to continuously monitor the lab air quality for a large spectrum of fumes and vapours. Halo Sense comes in 3 versions: Volatile Organic Compounds (VOC), acid and formaldehyde.
  • Simple communication by LED pulsation system
  • Power supplies in compliance with various international standards
  • RJ45 cable provided
  • Default settings: audible alarm disabled, visual alarm activated (both alarms can be disabled)
  • To be installed: on the ceiling, on the wall, on the bench, or under
When Halo senses fumes or vapours, a soft band of light pulses to let the user know that the air is polluted.
The user has only to download the Erlab eGuard App and it will send alerts to the preferred device. If the laboratory air is polluted, it allows the user to take action before pollution levels become a hazard.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description W×D×H Pk VWR Catalog Number Quantity
Halo Sense VOC, for laboratories with volatile organic compounds 175×165×64 mm 1 135-2277
Halo Sense Acid, for laboratories with acid vapours 175×165×64 mm 1 135-2278
Halo Sense Formaldehyde, for laboratories with formaldehyde vapours 175×165×64 mm 1 135-2279
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty