Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Centrifugal devices, Microsep™ Advance and Macrosep® Advance

Supplier: Pall Laboratory

Microsep™ Advance centrifugal devices

Precise, quick recovery of microlitre volumes of concentrate.

  • High recoveries, typically greater than 90%
  • With deadstop to prevent samples from spinning to dryness
  • Fits centrifuges that accept standard 17×100 mm tubes
Macrosep® Advance centrifugal devices

Quickly concentrates up to 20 ml of biological sample without valuable sample loss.

  • High recoveries, typically greater than 90%
  • With deadstop to prevent samples from spinning to dryness
  • Low protein binding Omega™ membrane and polypropylene housing minimise losses due to non specific binding
  • Fits centrifuges that accept standard 50 ml conical tubes
Applications for Microsep™ Advance centrifugal devices: Concentrate dilute protein samples prior to electrophoresis. Exchange buffer and remove salt in samples. Remove proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies. Isolate low molecular weight compounds from fermentation broths for natural product screening. Recover biomolecules from cell culture supernatants or lysates. Clarify samples with gross particulate.

Applications for Macrosep® Advance centrifugal devices: Concentrate and desalt proteins. Exchange buffer or remove salt of chromatography eluates and gradient fractions. Recover proteins or other molecules from cell culture supernatants. Remove particulate from aqueous solutions.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

Microsep™ Advance with Omega™ membrane
MWCO Colour code Pk VWR Catalog Number Quantity
1 kD Yellow 24 516-9920
1 kD Yellow 100 516-0485
3 kD Grey 24 516-0370
3 kD Grey 100 516-0371
10 kD Blue 24 516-0372
10 kD Blue 100 516-0373
30 kD Red 24 516-0374
30 kD Red 100 516-0375
100 kD Clear 24 516-0376
100 kD Clear 100 516-0377

Microsep™ Advance with Supor® membrane
Pore size Colour code Pk VWR Catalog Number Quantity
0,2 µm Turquoise 24 516-0378
0,2 µm Turquoise 100 516-0381
0,45 µm Purple 24 516-0382
0,45 µm Purple 100 516-0383

Macrosep® Advance with Omega™ membrane
MWCO Colour code Pk VWR Catalog Number Quantity
1 kD Yellow 6 516-0483
1 kD Yellow 24 516-9919
1 kD Yellow 100 516-0484
3 kD Grey 6 516-0354
3 kD Grey 24 516-0355
3 kD Grey 100 516-0356
10 kD Blue 6 516-0357
10 kD Blue 24 516-0358
10 kD Blue 100 516-0359
30 kD Red 6 516-0360
30 kD Red 24 516-0361
30 kD Red 100 516-0362
100 kD Clear 6 516-0363
100 kD Clear 24 516-0364
100 kD Clear 100 516-0365

Macrosep® Advance with Supor® membrane
Pore size Colour code Pk VWR Catalog Number Quantity
0,2 µm Turquoise 24 516-0366
0,2 µm Turquoise 100 516-0367
0,45 µm Purple 24 516-0368
0,45 µm Purple 100 516-0369
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Microsep™ Macrosep®
Filtration area (cm²) 3,3 7,2
Max. RCF (×g) 14000
Dead volume 40 µl 80 µl
Ø×H (mm) 17×120 50×120
Sample volume (µl) max. 5000 max. 20000
Temperature (°C) 0 - 40
pH range 1 - 14

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty