Complete ICP-MS standards range at 10 mg/l
68 single elements ICP-MS standards for metal trace analysis calibration
Large ICP-MS single element standards range at 10 mg/l delivered with an extensive Certificate of Analysis (NIST traceability, controlled by an accredited ISO 17025 lab, ISO guide 34). 
  • Produced from high purity acids, water ASTM 18 MΩ and salts 99,999% 
  • Solution assayed by titration
  • Final concentration verified against reference materials from NIST
  • Produced by accreditated supplier ISO 17025 and ISO guide 34
  • Complete Certificate of Analysis delivered with the product in a individual plastic bag
  Description Pk Cat. No. Promo price Quantity
  Aluminium standard solutions for ICP-MS, ARISTAR® 100ml 85548.180E    
  Arsenic standard solutions for ICP-MS, ARISTAR® 100 ml 85549.180E  
  Antimony standard solutions for ICP-MS, ARISTAR® 100 ml 85595.180E  
  Boron standard solutions for ICP-MS, ARISTAR® 100 ml 85551.180E  
  Barium standard solutions for ICP-MS, ARISTAR® 100 ml 85552.180E  
  Beryllium standard solutions for ICP-MS, ARISTAR® 100 ml 85553.180E  
  Bismuth standard solutions for ICP-MS, ARISTAR® 100 ml 85554.180E  
  Calcium standard solutions for ICP-MS, ARISTAR® 100 ml 85555.180E  
  Cadmium standard solutions for ICP-MS, ARISTAR® 100 ml 85556.180E  
  Cerium standard solutions for ICP-MS, ARISTAR® 100 ml 85557.180E  
  Cobalt standard solutions for ICP-MS, ARISTAR® 100 ml 85558.180E  
  Chromium standard solutions for ICP-MS, ARISTAR® 100 ml 85559.180E  
  Cesium standard solutions for ICP-MS, ARISTAR® 100 ml 85560.180E  
  Copper standard solutions for ICP-MS, ARISTAR® 100 ml 85561.180E  
  Dysprosium standard solutions for ICP-MS, ARISTAR® 100 ml 85562.180E  
  Erbium standard solutions for ICP-MS, ARISTAR® 100 ml 85563.180E  
  Europium standard solutions for ICP-MS, ARISTAR® 100 ml 85564.180E  
  Gallium standard solutions for ICP-MS, ARISTAR® 100 ml 85566.180E  
  Gadolinium standard solutions for ICP-MS, ARISTAR® 100 ml 85567.180E  
  Germanium standard solutions for ICP-MS, ARISTAR® 100 ml 85568.180E  
  Gold standard solutions for ICP-MS, ARISTAR® 100 ml 85550.180E  
  Hafnium standard solutions for ICP-MS, ARISTAR® 100 ml 85569.180E  
  Holmium standard solutions for ICP-MS, ARISTAR® 100 ml 85571.180E  
  Iron standard solutions for ICP-MS, ARISTAR® 100 ml 85565.180E  
  Indium standard solutions for ICP-MS, ARISTAR® 100 ml 85572.180E  
  Iridium standard solutions for ICP-MS, ARISTAR® 100 ml 85573.180E  
  Lanthanum standard solutions for ICP-MS, ARISTAR® 100 ml 85575.180E  
  Lithium standard solutions for ICP-MS, ARISTAR® 100 ml 85576.180E  
  Lutetium standard solutions for ICP-MS, ARISTAR® 100 ml 85577.180E  
  Lutetium standard solutions for ICP-MS, ARISTAR® 100 ml 85579.180E  
  Lead standard solutions for ICP-MS, ARISTAR® 100 ml 85586.180E  
  Magnesium standard solutions for ICP-MS, ARISTAR® 100 ml 85578.180E  
  Mercury standard solutions for ICP-MS, ARISTAR® 100 ml 85570.180E  
  Molybdenum standard solutions for ICP-MS, ARISTAR® 100 ml 85580.180E  
  Niobium standard solutions for ICP-MS, ARISTAR® 100 ml 85582.180E  
  Neodymium standard solutions for ICP-MS, ARISTAR® 100 ml 85583.180E  
  Nickel standard solutions for ICP-MS, ARISTAR® 100 ml 85584.180E  
  Palladium standard solutions for ICP-MS, ARISTAR® 100 ml 85587.180E  
  Praseodymium standard solutions for ICP-MS, ARISTAR® 100 ml 85588.180E  
  Platinum standard solutions for ICP-MS, ARISTAR® 100 ml 85589.180E  
  Phosphorus standard solutions for ICP-MS, ARISTAR® 100 ml 85585.180E  
  Potassium standard solutions for ICP-MS, ARISTAR® 100 ml 85574.180E  
  Rubidium standard solutions for ICP-MS, ARISTAR® 100 ml 85590.180E  
  Rhenium standard solutions for ICP-MS, ARISTAR® 100 ml 85591.180E  
  Rhodium standard solutions for ICP-MS, ARISTAR® 100 ml 85592.180E  
  Ruthenium standard solutions for ICP-MS, ARISTAR® 100 ml 85593.180E  
  Silver standard solutions for ICP-MS, ARISTAR® 100 ml 85547.180E  
  Sodium standard solutions for ICP-MS, ARISTAR® 100 ml 85581.180E  
  Scandium standard solutions for ICP-MS, ARISTAR® 100 ml 85596.180E  
  Selenium standard solutions for ICP-MS, ARISTAR® 100 ml 85597.180E  
  Silicon standard solutions for ICP-MS, ARISTAR® 100 ml 85598.180E  
  Samarium standard solutions for ICP-MS, ARISTAR® 100 ml 85599.180E  
  Strontium standard solutions for ICP-MS, ARISTAR® 100 ml 85631.180E  
  Sulphur standard solutions for ICP-MS, ARISTAR® 100 ml 85594.180E  
  Tantalum standard solutions for ICP-MS, ARISTAR® 100 ml 85632.180E  
  Terbium standard solutions for ICP-MS, ARISTAR® 100 ml 85633.180E  
  Tellurium standard solutions for ICP-MS, ARISTAR® 100 ml 85634.180E  
  Titanium standard solutions for ICP-MS, ARISTAR® 100 ml 85636.180E  
  Thallium standard solutions for ICP-MS, ARISTAR® 100 ml 85639.180E  
  Thulium standard solutions for ICP-MS, ARISTAR® 100 ml 85640.180E  
  Tin standard solutions for ICP-MS, ARISTAR® 100 ml 85630.180E  
  Tungsten standard solutions for ICP-MS, ARISTAR® 100 ml 85644.180E  
  Uranium standard solutions for ICP-MS, ARISTAR® 100 ml 85641.180E  
  Vanadium standard solutions for ICP-MS, ARISTAR® 100 ml 85643.180E  
  Vanadium standard solutions for ICP-MS, ARISTAR® 100 ml 85646.180E  
  Ytterbium standard solutions for ICP-MS, ARISTAR® 100 ml 85647.180E  
  Zinc standard solutions for ICP-MS, ARISTAR® 100 ml 85648.180E  
  Zirconium standard solutions for ICP-MS, ARISTAR® 100 ml 85649.180E