VWR Collection cleanroom nitrile gloves
NEW!
Switch cleanroom gloves from latex to nitrile with the brand new gloves from VWR Collection.
  • High tensile strength and strong permeation resistance against a wide variety of solvents, acids and bases
  • Double bagged in sealed polyethylene cleanroom bags (100 pieces/liner; 10 liners in a bag/carton). Sterile gloves individually packed (50 pairs in a liner, 4 liners in a bag/carton)
  • Extractables measured according to IEST-RP-CC005.3
  • Sterile version: Endotoxin level <20 EU/glove
  • Compliant to the PPE Directive (89/686/EEC) and EN 374
  • Contain no natural rubber latex