Secondary reference standards for Gas Chromatography
Wide range of 24 different GC secondary reference standards for various chromatographic and analytical applications. FDA, USP and EP, all recognise the use of secondary standards and working standards that are established with reference to the corresponding primary standard.
  • Purity and specific impurity profile determined by GC-MS (reported in the Certificate of Analysis)
  • Assay over 99,9% 
  • Cost effective alternative to the pharmacopeias primary standards
  • Available in 100 ml glass bottle
  • Traceability data to specific current lot of primary standard
  Description Pk Cat. No. Promo price Quantity
  Acetone standard for GC 100 ml 85653.180E  
  Acetonitrile standard for GC 100 ml 85654.180E  
  Butyl acetate standard for GC 100 ml 85655.180E  
  Cyclohexane standard for GC 100 ml 85656.180E  
  1,2 Dichloroethane standard for GC 100 ml 85657.180E  
  Dichlormethane (DCM), stabilised standard for GC 100 ml 85658.180E  
  Di-isopropyl ether, stabilised standard for GC 100 ml 85659.180E  
  N,N-Dimethylacetamide standard for GC 100 ml 85661.180E  
  N,N-Dimethylformamide standard for GC 100 ml 85662.180E  
  Dimethylsulphoxide (DMSO) standard for GC 100 ml 85663.180E  
  1,4-Dioxane, stabilised standard for GC 100 ml 85664.180E  
  Ethanol standard for GC 100 ml 85665.180E  
  Ethyl acetate standard for GC 100 ml 85666.180E  
  n-Heptane standard for GC 100 ml 85667.180E  
  n-Hexane standard for GC 100 ml 85668.180E  
  iso-Butyl methyl ketone standard for GC 100 ml 85673.180E  
  Methanol standard for GC 100 ml 85670.180E  
  Methyl ethyl ketone (butanone) standard for GC 100 ml 85672.180E  
  1-Propanol standard for GC 100 ml 85675.180E  
  2-Propanol standard for GC 100 ml 85676.180E  
  tert-Butyl methyl ether standard for GC 100 ml 85671.180E  
  2,2,4-Trimethylpentane standard for GC 100 ml 85674.180E  
  Tetrahydrofuran (THF) standard for GC 100 ml 85677.180E  
  Toluene standard for GC 100 ml 85678.180E