BOD Measurement/Respiration
Biological Oxygen Demand (BOD) is an important measure of quality in water resource management. VWR offers products for the different methods that are available.
BOD incubators
All BOD methods require samples to be kept at 20 °C (68 °F) for 5 days. VWR offers a wide range of temperature-controlled incubators
Respiration BOD: BOD self-checks
A reduction in oxygen causes a definite pressure difference that is measured by a pressure sensor. Practical and easy to perform!
Dilution BOD
BOD measurement according to EN 1899...
BOD Photometric Method
The BOD test is a photometric cell test from Merck...