Oxivir Plus - powerful detergent and broad spectrum disinfectant
Single step cleaning and disinfection, with no compromise on efficacy, thanks to the AHP (Accelerated Hydrogen Peroxide) technology.
- Efficient descaling detergent, streak-free and scale retardant
- Broad spectrum disinfectant with fast action on bacteria, fungi, yeast and viruses
- Environmentally-friendly: Hydrogen peroxide naturally degrades into water and oxygen
- Safe for the staff: Ready to use solution is non hazardous and has low VOCs
- Compatible for use on water- and acid-resistant surfaces, as well as on fabrics
- Available in a ready to use 750 ml spray bottle or as a concentrate in a 5 l canister