Density standards
182 density standards for use at different temperatures (15°C, 20°C, 25°C, 40°C, 50°C, 60°, 80°C)
Extensive range of density standards in accordance with ASTM D1480-12 for density or relative density (specific gravity) measurements  by Bingham Pycnometer.
  • High accuracy
  • Verified using Bingham Pycnometer
  • All products tested in accordance with ASTM D1480-12 guidelines
  • Consistent, traceable, certified
  • Certificates of Analysis and Safety Data Sheets available online
  Description Pk Cat. No. Promo price Quantity
  Density standard, 0,6960 g/ml at 15 °C 100 ml 85140.180E  
  Density standard, 0,7073 g/ml at 15 °C 100 ml 85141.180E  
  Density standard, 0,7184 g/ml at 15 °C 100 ml 85142.180E  
  Density standard, 0,7298 g/ml at 15 °C 100 ml 85143.180E  
  Density standard, 0,7411 g/ml at 15 °C 100 ml 85144.180E  
  Density standard, 0,7524 g/ml at 15 °C 100 ml 85145.180E  
  Density standard, 0,7721 g/ml at 15 °C 100 ml 85146.180E  
  Density standard, 0,7933 g/ml at 15 °C 100 ml 85147.180E  
  Density standard, 0,8168 g/ml at 15 °C 100 ml 85148.180E  
  Density standard, 0,8428 g/ml at 15 °C 100 ml 85149.180E  
  Density standard, 0,8715 g/ml at 15 °C 100 ml 85150.180E  
  Density standard, 0,9135 g/ml at 15 °C 100 ml 85151,180E  
  Density standard, 0,9514 g/ml at 15 °C 100 ml 85152.180E  
  Density standard, 1,0040 g/ml at 15 °C 100 ml 85153.180E  
  Density standard, 1,0337 g/ml at 15 °C 100 ml 85154.180E  
  Density standard, 1,0828 g/ml at 15 °C 100 ml 85155.180E  
  Density standard, 1,1661 g/ml at 15 °C 100 ml 85156.180E  
  Density standard, 1,3318 g/ml at 15 °C 100 ml 85158.180E  
  Density standard, 1,5820 g/ml at 15 °C 100 ml 85160.180E  
  Density standard, 1,7495 g/ml at 15 °C 100 ml 85161.180E  
  Density standard, 2,0846 g/ml at 15 °C 100 ml 85163.180E  
  Density standard, 2,2568 g/ml at 15 °C 100 ml 85164.180E  
  Density standard, 2,4261 g/ml at 15 °C 100 ml 85165.180E  
  Density standard, 2,6055 g/ml at 15 °C 100 ml 85166.180E  
  Density standard, 2,7588 g/ml at 15 °C 100 ml 85167.180E  
  Density standard, 2,9418 g/ml at 15 °C 100 ml 85168.180E  
  Density standard, 3.1140 g/ml at 15 °C 100 ml 85169.180E