Temperature: measurement - logging – accredited services
In addition to an extensive range of devices for measuring or logging temperature, VWR also offers a varied range of services for monitoring and accredited calibration.
See below for further information and product selection.
For a complete overview of all temperature humidity applications, from strips to advanced dataloggers; download the HSG.
Product Selection
Just measuring temperature, -50°C to +70°C
  • With probe, bottle probe or ambient
  • Readability 0.1°C, Accuracy +/- 1°C
Measuring temperature , with alarm, -50°C to +70°C
  • With probe, bottle probe or ambient
  • Readability 0.1°C, Accuracy +/- 1°C
Measuring temperature, with radio signal, -30°C to +70°C
  • With bottle probe
  • Readability 0.1°C, Accuracy +/- 1°C
High accuracy temperature measurement, with alarm, -50°C to +70°C
  • With probes, bottle probes
  • Readability 0.01°C, Accuracy +/-0.1°C
Temperature Datalogging, ideal for transport, -35°C to +50°C
  • Without probe
  • Readability 0.1°C, Accuracy +/- 0.2°C
  • With cradle&cable or blue tooth
High accuracy Temperature Datalogging, -50°C to +70°C
  • With probes, bottle probes
  • Readability 0.01°C, Accuracy +/-0.25°C
  • Up to 1.000.000 data memory
High accuracy, Temperature Datalogging, -90°C to +105°C
  • With probes
  • Readability 0.01°C, Accuracy +/-0.2°C and 0.1°C
  • Up to 1.000.000 data memory
High accuracy Temperature Datalogging, -200°C to +105°C
  • With probes
  • Readability 0.01°C, Accuracy +/-0.2°C and 0.1°C
  • Up to 1.000.000 data memory
Wireless temperature (& more) Datalogging, Sense Anywhere,-200°C to + 200°C
  • Without probes
  • Readability 0.01°C, Accuracy +/- 0.15°C
  • Real time, Cloud based
Accredited calibration for all your thermometers and dataloggers
Brand independent ISO17025-accredited calibration services for temperature-devices like thermometers, dataloggers, blocks and baths.
Learn more
Your advantages
  • Approved quality
  • Detailed Proces-documentation
  • All brands & models
  • NIST traceable
  • ISO 17025 calibration-certificate
  • Due time reminder system
  • Combine with humidity-calibration