Dew-point and humidity meters
For certain specific applications in the pharmaceutical industry, the calibration must be carried out every 3 months, as measuring devices are subject to the validation process and even the smallest fluctuations can have a large influence on the application.
Humidity calibrator
  • For calibration of humidity sensors, transmitters and portable devices
  • Generates stable humidity values of 10 to 90% RH
  • Compact and easy to transport
  • Rapid stabilisation of the test chamber
  • Optional: independent traceable reference hygrometer
  • Several sensors can be calibrated simultaneously
Humidity / temperature calibrator
  • For calibration of rF humidity sensors and transmitters
  • Temperatures from 10 to 50 °C with a precision of 0.1 °C and humidity of 10 to 90% RH
  • Precise and repeatable calibrations are possible
  • Generates humidity and temperature simultaneously
  • Excellent stability
  • Compact and easy to transport
  • Optional: built-in data recorder, logs the built-in reference sensor and the sensors to be calibrated
  • Suitable for sensors with Ø betw. 5 and 25 mm
Contact us