Would you like to log in again?
New J.T.Baker® Direct Dispense packaging
AVOID CLUMPING
New J.T.Baker® Direct Dispense packaging
Keep salts, buffers and cell culture products free-flowing
  • For any powder material
  • Eliminate time-consuming de-clumping and subdivision steps
  • Available in 3 sizes: 30, 65 and 121 l
  • Compatible with traditional and single-use manufacturing equipment
  • Closed system lowers your risk of cross-contamination
Direct Dispense packaging keeps your material flowing smoothly. For easy and controlled delivery directly to your process.