Sample Preparation for Life Science
Centrifugal Devices
Ensure rapid processing of samples with typical recoveries greater than 90%.
Nanosep®
Simple, reliable concentrating and desalting of 50 to 500 µl samples.
Microsep™ Advance
Confidence in rapid recovery of <100 µl volumes of concentrate from starting volumes up to 5 ml.
Macrosep® Advance
Quickly concentrates up to 20 ml of biological sample without valuable sample loss.
Application
  • For use of proteins and nucleic acids
  • Concentration
  • Buffer exchange
  • De-salting
  • Fractionation
Acroprep Advance Filter Plates
For high throughput sample prep and detection procedures
  • Provides consistency in filtration times, as well as efficient sample and bead recovery
  • Available in a variety of membrane configurations, plate colours, well volumes, and outlet tip lengths
  • Plates are constructed from chemically-resistant, biologically-inert polypropylene
  • Serialised barcode label allows for use in automated tracking systems
Applications
  • Concentration, purification, and desalting of proteins and peptides
  • Bead-/resin-based applications
  • Gross fractionation and lysate clarification
  • pDNA, Gdna and local RNA purification
  • General filtration
Flip Catalogue